accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 631-71-0

IUPAC Name: ethyl (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylate
Synonyms: ethyl (1R,4aR,4bR,10aR)-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylate
Molecular Weight (g/mol): 330.51
Molecular Formula: C22H34O2
InChi Key: AGUBCDYYAKENKG-YVNJGZBMSA-N
SMILES: CCOC(=O)[C@]1(C)CCC[C@]2(C)[C@H]3CCC(=CC3=CC[C@@H]12)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results