accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSCl

CAS RN 6310-09-4

IUPAC Name: 1-(5-chlorothiophen-2-yl)ethan-1-one
Synonyms: 1-(5-chlorothiophen-2-yl)ethanone
Molecular Weight (g/mol): 160.62
Molecular Formula: C6H5ClOS
InChi Key: HTZGPEHWQCRXGZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)S1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)