accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H2N

CAS RN 6310-21-0

IUPAC Name: 2-tert-butylaniline
Synonyms: 2-tert-butylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: AEIOZWYBDBVCGW-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC=C1N
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)