accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63104-44-9

IUPAC Name: 1,3-dimethyl 2,2-bis(prop-2-yn-1-yl)propanedioate
Synonyms: 1,3-dimethyl 2,2-bis(prop-2-yn-1-yl)propanedioate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: AKISDSWMEPTCRL-UHFFFAOYSA-N
SMILES: COC(=O)C(CC#C)(CC#C)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)