accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BrOHO

CAS RN 6311-37-1

IUPAC Name: 4-amino-3-bromobenzoic acid
Synonyms: 4-amino-3-bromobenzoic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: BFIVZIVVJNFTIQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Br)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results