accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6313-33-3

IUPAC Name: methanimidamide hydrochloridyl
Synonyms: formamidine hydrochloridyl
Molecular Weight (g/mol): 79.51
Molecular Formula: CH4ClN2
InChi Key: OEJWQRAWUVWBIE-UHFFFAOYSA-N
SMILES: [Cl].NC=N
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)