accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63134-04-3

IUPAC Name: N-[3-(ethylamino)-4-methylphenyl]acetamide
Synonyms: N-[3-(ethylamino)-4-methylphenyl]acetamide
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: MRNDZGBBVKUGJY-UHFFFAOYSA-N
SMILES: CCNC1=CC(NC(C)=O)=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results