CAS RN 6314-28-9
IUPAC Name:
1-benzothiophene-2-carboxylic acid
Synonyms:
1-benzothiophene-2-carboxylic acid
Molecular Weight (g/mol):
178.21
Molecular Formula:
C9H6O2S
InChi Key:
DYSJMQABFPKAQM-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC2=CC=CC=C2S1