accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6314-43-8

IUPAC Name: 1-(1-benzothiophen-2-yl)methanamine
Synonyms: 1-(1-benzothiophen-2-yl)methanamine
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: WKYFWFHTABURGB-UHFFFAOYSA-N
SMILES: NCC1=CC2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results