accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63148-56-1

IUPAC Name: methyl(3,3,3-trifluoropropyl)silanone
Synonyms: methyl(3,3,3-trifluoropropyl)silanone
Molecular Weight (g/mol): 156.18
Molecular Formula: C4H7F3OSi
InChi Key: CUMCLKHSZZGALD-UHFFFAOYSA-N
SMILES: C[Si](=O)CCC(F)(F)F
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)