accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOOHOHOSOOHOH2ONaNa

CAS RN 63148-74-3

IUPAC Name: bis(hydroxymethanesulfonic acid) hydrate disodium
Synonyms: bis(hydroxymethanesulfonic acid) hydrate disodium
Molecular Weight (g/mol): 288.19
Molecular Formula: C2H10Na2O9S2
InChi Key: ZOOZASWIMKXRIF-UHFFFAOYSA-N
SMILES: O.[Na].[Na].OCS(O)(=O)=O.OCS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)