accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6317-18-6

IUPAC Name: {[(cyanosulfanyl)methyl]sulfanyl}carbonitrile
Synonyms: methylene bisthiocyanate
Molecular Weight (g/mol): 130.18
Molecular Formula: C3H2N2S2
InChi Key: JWZXKXIUSSIAMR-UHFFFAOYSA-N
SMILES: N#CSCSC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results