accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 6317-73-3

IUPAC Name: (4-phenoxyphenyl)(phenyl)methanone
Synonyms: (4-phenoxyphenyl)(phenyl)methanone
Molecular Weight (g/mol): 274.32
Molecular Formula: C19H14O2
InChi Key: ITVUPWDTDWMACZ-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C(OC2=CC=CC=C2)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results