CAS RN 6318-51-0
IUPAC Name:
2-(4-chlorobenzoyl)pyridine
Synonyms:
2-(4-chlorobenzoyl)pyridine
Molecular Weight (g/mol):
217.65
Molecular Formula:
C12H8ClNO
InChi Key:
KHXSJSBQIWAIEG-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1
1
–
1
of
1
results
1
2-(4-Chlorobenzoyl)pyridine 97.0+%, TCI America™
CAS: 6318-51-0 Molecular Formula: C12H8ClNO Molecular Weight (g/mol): 217.65 MDL Number: MFCD02930888 InChI Key: KHXSJSBQIWAIEG-UHFFFAOYSA-N Synonym: 4-Chlorophenyl 2-Pyridyl Ketone PubChem CID: 80594 IUPAC Name: 2-(4-chlorobenzoyl)pyridine SMILES: ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1