accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6319-40-0

IUPAC Name: 4-bromo-3-nitrobenzoic acid
Synonyms: benzoic acid, 4-bromo-3-nitro-
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: RVCTZJVBWNFYRU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)