accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2BOH3CH3CH3CCH3O

CAS RN 631911-01-8

IUPAC Name: 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 233.12
Molecular Formula: C13H20BNO2
InChi Key: ZZMUAVWNSFFTLI-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results