accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHOO

CAS RN 632-46-2

IUPAC Name: 2,6-dimethylbenzoic acid
Synonyms: 2,6-dimethylbenzoic acid
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: HCBHQDKBSKYGCK-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)