accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3NHH3COOCH3CH3

CAS RN 632-93-9

IUPAC Name: 3,5-diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: 3,5-diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol): 267.33
Molecular Formula: C14H21NO4
InChi Key: CDVAIHNNWWJFJW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)NC(C)=C(C1C)C(=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)