accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6320-03-2

IUPAC Name: 2-chlorobenzene-1-thiol
Synonyms: benzenethiol, 2-chloro-
Molecular Weight (g/mol): 144.62
Molecular Formula: C6H5ClS
InChi Key: PWOBDMNCYMQTCE-UHFFFAOYSA-N
SMILES: SC1=CC=CC=C1Cl
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)