accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6320-40-7

IUPAC Name: 1,3,5-tribromo-2-methylbenzene
Synonyms: 2,4,6-tribromotoluene
Molecular Weight (g/mol): 328.83
Molecular Formula: C7H5Br3
InChi Key: BFRIZWKDNUHPHL-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results