accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6320-96-3

IUPAC Name: 3-bromopropanamide
Synonyms: 3-bromopropanamide
Molecular Weight (g/mol): 151.99
Molecular Formula: C3H6BrNO
InChi Key: DBIVLAVBOICUQX-UHFFFAOYSA-N
SMILES: NC(=O)CCBr
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results