accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63212-53-3

IUPAC Name: 2-chloro-3-ethyl-1,3-benzoxazol-3-ium; tetrafluoroboranuide
Synonyms: 2-chloro-3-ethyl-1,3-benzoxazol-3-ium tetrafluoroborate
Molecular Weight (g/mol): 269.43
Molecular Formula: C9H9BClF4NO
InChi Key: MLQZANYFUYLJRK-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC[N+]1=C(Cl)OC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results