accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOHN(S)(R)OO

CAS RN 63216-49-9

IUPAC Name: (1R,2S)-2-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylate
Synonyms: (1R,2S)-2-[(tert-butoxycarbonyl)amino]cyclohexane-1-carboxylate
Molecular Weight (g/mol): 242.3
Molecular Formula: C12H20NO4
InChi Key: QJEQJDJFJWWURK-BDAKNGLRSA-M
SMILES: CC(C)(C)OC(=O)N[C@H]1CCCC[C@H]1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)