accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6323-18-8

IUPAC Name: 1-(2-chlorophenyl)propan-1-one
Synonyms: 1-(2-chlorophenyl)propan-1-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: BTSCBJDORATYKJ-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results