accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63234-80-0

IUPAC Name: 3-(2-chloroethyl)-2-methyl-4H,6H,7H,8H,9H-pyrido[1,2-a]pyrimidin-4-one
Synonyms: 3-(2-chloroethyl)-2-methyl-6H,7H,8H,9H-pyrido[1,2-a]pyrimidin-4-one
Molecular Weight (g/mol): 226.7
Molecular Formula: C11H15ClN2O
InChi Key: CMWCQQUYLPYOMY-UHFFFAOYSA-N
SMILES: CC1=C(CCCl)C(=O)N2CCCCC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results