accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6326-27-8

IUPAC Name: hydrogen 4-methylbenzene-1-carboximidamide chloride
Synonyms: hydrogen 4-methylbenzenecarboximidamide chloride
Molecular Weight (g/mol): 170.64
Molecular Formula: C8H11ClN2
InChi Key: MTDUPZAXNYELOU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(C=C1)C(N)=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results