accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOHN

CAS RN 6326-79-0

IUPAC Name: 6-bromo-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 6-bromo-1H-indole-2,3-dione
Molecular Weight (g/mol): 226.03
Molecular Formula: C8H4BrNO2
InChi Key: HVPQMLZLINVIHW-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(NC(=O)C2=O)=C1
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)