accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6326-83-6

IUPAC Name: 2-({[(carboxymethyl)sulfanyl]methanethioyl}sulfanyl)acetic acid
Synonyms: ({[(carboxymethyl)sulfanyl]methanethioyl}sulfanyl)acetic acid
Molecular Weight (g/mol): 226.28
Molecular Formula: C5H6O4S3
InChi Key: GQECANUIPBFPLA-UHFFFAOYSA-N
SMILES: OC(=O)CSC(=S)SCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)