CAS RN 6327-59-9
CAS RN 6327-59-9
IUPAC Name:
1,4-dibenzyl 2-aminobutanedioate hydrochloridyl
Synonyms:
1,4-dibenzyl 2-aminobutanedioate hydrochloridyl
Molecular Weight (g/mol):
348.8
Molecular Formula:
C18H19ClNO4
InChi Key:
USJBDTOKIAONQV-UHFFFAOYNA-N
SMILES:
[Cl].NC(CC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
H-Asp OBzl -OBzl.HCl, Advanced ChemTech
CAS: 6327-59-9 Molecular Formula: C18H19ClNO4 Molecular Weight (g/mol): 348.80 InChI Key: USJBDTOKIAONQV-UHFFFAOYNA-N IUPAC Name: 1,4-dibenzyl 2-aminobutanedioate hydrochloridyl SMILES: [Cl].NC(CC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1