accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 633-12-5

IUPAC Name: 2,4,6-tribromobenzoic acid
Synonyms: benzoic acid, 2,4,6-tribromo-
Molecular Weight (g/mol): 358.81
Molecular Formula: C7H3Br3O2
InChi Key: GHVMJSHEGZYRQL-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C=C(Br)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results