accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNHSOOONa(E/Z)

CAS RN 633-96-5

IUPAC Name: sodium 4-[2-(2-oxo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]benzene-1-sulfonate
Synonyms: acid orange 7
Molecular Weight (g/mol): 350.32
Molecular Formula: C16H11N2NaO4S
InChi Key: IGQFKZCLTLAWLO-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(NN=C2C(=O)C=CC3=CC=CC=C23)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (1)
Quantity 
  • (4)
  • (3)
  • (2)
Grade 
  • (1)
  • (2)
  • (4)
Color 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)