accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6330-51-4

IUPAC Name: N-(naphthalen-1-yl)formamide
Synonyms: N-(naphthalen-1-yl)formamide
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: CGRYTQQVSFZYCI-UHFFFAOYSA-N
SMILES: O=CNC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results