accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6331-09-5

IUPAC Name: ethyl 2-aminopropanoate hydrochloridyl
Synonyms: ethyl 2-aminopropanoate hydrochloridyl
Molecular Weight (g/mol): 152.6
Molecular Formula: C5H11ClNO2
InChi Key: CYHRNDXVTAWKMO-UHFFFAOYNA-N
SMILES: [Cl].CCOC(=O)C(C)N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)