accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH2OOO

CAS RN 6331-61-9

IUPAC Name: 2-[2-methoxy-4-(prop-2-en-1-yl)phenoxy]acetate
Synonyms: 2-[2-methoxy-4-(prop-2-en-1-yl)phenoxy]acetate
Molecular Weight (g/mol): 221.23
Molecular Formula: C12H13O4
InChi Key: FQBPCNCFOCPCJP-UHFFFAOYSA-M
SMILES: COC1=CC(CC=C)=CC=C1OCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results