CAS RN 6334-25-4
IUPAC Name:
N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide
Synonyms:
N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide
Molecular Weight (g/mol):
320.39
Molecular Formula:
C14H28N2O6
InChi Key:
OKRNLSUTBJUVKA-UHFFFAOYSA-N
SMILES:
OCCN(CCO)C(=O)CCCCC(=O)N(CCO)CCO
1
–
1
of
1
results
1
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide 98.0+%, TCI America™
CAS: 6334-25-4 Molecular Formula: C14H28N2O6 Molecular Weight (g/mol): 320.39 MDL Number: MFCD07787618 InChI Key: OKRNLSUTBJUVKA-UHFFFAOYSA-N PubChem CID: 95283 IUPAC Name: N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide SMILES: OCCN(CCO)C(=O)CCCCC(=O)N(CCO)CCO