accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6336-45-4

IUPAC Name: dibutyl ethenylboronate
Synonyms: dibutyl ethenylboronate
Molecular Weight (g/mol): 184.09
Molecular Formula: C10H21BO2
InChi Key: JQKJEPJLCSCBGW-UHFFFAOYSA-N
SMILES: CCCCOB(OCCCC)C=C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)