CAS RN 634-62-8
CAS RN 634-62-8
IUPAC Name:
hydrazine
Synonyms:
hydrazine diamine (specific compound) hydrazine base salts of hydrazine hydrazine, its derivatives and their salts anhydrous hydrazine hydrazine sulfate (product) nitrogen hydride diamine ccris 335
Molecular Weight (g/mol):
32.05
Molecular Formula:
H4N2
InChi Key:
OAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILES:
NN
1
–
1
of
1
results
1
Hydrazine L-(+)-Tartrate 98.0+%, TCI America™

CAS: 634-62-8 Molecular Formula: C4H10N2O6 Molecular Weight (g/mol): 182.132 MDL Number: MFCD00070610 InChI Key: GHPDWAAGDJPBLL-ZVGUSBNCSA-N PubChem CID: 44630322 IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid;hydrazine SMILES: C(C(C(=O)O)O)(C(=O)O)O.NN