accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 634-66-2

IUPAC Name: 1,2,3,4-tetrachlorobenzene
Synonyms: 1,2,3,4-tetrachlorobenzene caswell no. 825 epa pesticide chemical code 061101
Molecular Weight (g/mol): 215.88
Molecular Formula: C6H2Cl4
InChi Key: GBDZXPJXOMHESU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)