accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 634-67-3

IUPAC Name: 2,3,4-trichloroaniline
Synonyms: 2,3,4-trichloroaniline
Molecular Weight (g/mol): 196.46
Molecular Formula: C6H4Cl3N
InChi Key: RRJUYQOFOMFVQS-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(Cl)=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results