accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63417-81-2

IUPAC Name: 5-(chloromethyl)-3-(thiophen-2-yl)-1,2,4-oxadiazole
Synonyms: 5-(chloromethyl)-3-(thiophen-2-yl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 200.64
Molecular Formula: C7H5ClN2OS
InChi Key: YOUDLOUFERNGRO-UHFFFAOYSA-N
SMILES: ClCC1=NC(=NO1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results