accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOHN(R)OOOO

CAS RN 63422-71-9

IUPAC Name: (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetate
Synonyms: (2R)-2-(4-ethyl-2,3-dioxopiperazine-1-carbonylamino)-2-phenylacetate
Molecular Weight (g/mol): 318.31
Molecular Formula: C15H16N3O5
InChi Key: JQEHQELQPPKXRR-LLVKDONJSA-M
SMILES: CCN1CCN(C(=O)N[C@@H](C([O-])=O)C2=CC=CC=C2)C(=O)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results