accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6343-27-7

IUPAC Name: 1-benzoylpyrrolidine-2,5-dione
Synonyms: 1-benzoylpyrrolidine-2,5-dione
Molecular Weight (g/mol): 203.2
Molecular Formula: C11H9NO3
InChi Key: CLFHABXQJQAYEF-UHFFFAOYSA-N
SMILES: O=C(N1C(=O)CCC1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results