accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6345-27-3

IUPAC Name: pyridine-4-carboximidamide hydrochloride
Synonyms: isonicotinamidine hydrochloride
Molecular Weight (g/mol): 157.6
Molecular Formula: C6H8ClN3
InChi Key: IONKMFGAXKCLMI-UHFFFAOYSA-N
SMILES: Cl.NC(=N)C1=CC=NC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)