accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH2CHOCH2H2COOCH2

CAS RN 63450-15-7

IUPAC Name: [(ethenyloxy)methoxy]ethene; ethenol; ethenyl acetate
Synonyms: formvar
Molecular Weight (g/mol): 230.26
Molecular Formula: C11H18O5
InChi Key: CCGLZRNKJSJOPC-UHFFFAOYSA-N
SMILES: OC=C.CC(=O)OC=C.C=COCOC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results