accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63467-30-1

IUPAC Name: 3-{iminiumyl[2-(propan-2-ylidene)hydrazin-1-yl]methyl}-1-phenylthiourea
Synonyms: 3-{iminio[2-(propan-2-ylidene)hydrazin-1-yl]methyl}-1-phenylthiourea
Molecular Weight (g/mol): 250.34
Molecular Formula: C11H16N5S
InChi Key: FWFDSNTZHZXYAD-UHFFFAOYSA-O
SMILES: CC(C)=NNC(=[NH2+])NC(=S)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results