CAS RN 634911-80-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Caspase Inhibitor III
The Caspase Inhibitor III, also referenced under CAS 634911-80-1, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.