accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H2N

CAS RN 63493-28-7

IUPAC Name: pentan-2-amine
Synonyms: 2-pentanamine
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: IGEIPFLJVCPEKU-UHFFFAOYNA-N
SMILES: CCCC(C)N
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)