accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3CO

CAS RN 635-67-6

IUPAC Name: 2-(acetyloxy)phenyl acetate
Synonyms: 1,2-benzenediol, diacetate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: FBSAITBEAPNWJG-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1OC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results