accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClO

CAS RN 635-84-7

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}-2-chloroethan-1-one
Synonyms: 1-{[1,1'-biphenyl]-4-yl}-2-chloroethanone
Molecular Weight (g/mol): 230.69
Molecular Formula: C14H11ClO
InChi Key: IQEIGQFNDLINOT-UHFFFAOYSA-N
SMILES: ClCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results