accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFCl

CAS RN 635305-46-3

IUPAC Name: 2-(3-chloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3-chloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 256.51
Molecular Formula: C12H15BClFO2
InChi Key: PFRXRUVRRFJIBF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results